Product Details
- SNP ID
-
rs138152773
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:54999312 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGGCACCTTCTATGCCATGGGCAG[C/T]GTCATGGTGATCGGGGGCATCATCT
- Phenotype
-
MIM: 606412
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BSND
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34561376] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BSND
- Gene Name
- barttin CLCNK type accessory beta subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_057176.2 |
369 |
Silent Mutation |
AGC,AGT |
S42S |
NP_476517.1 |
- Gene
- TMEM61
- Gene Name
- transmembrane protein 61
There are no transcripts associated with this gene.
View Full Product Details