Product Details

SNP ID
rs138160308
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:1373873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTTTAGCCCCGCCTTCAGCCA[C/G]ATGCGCCTCAGGTCTTTCTCGAACT
Phenotype
MIM: 609183
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AURKAIP1 PubMed Links

Gene Details

Gene
AURKAIP1
Gene Name
aurora kinase A interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127229.1 702 Missense Mutation ATC,ATG I176M NP_001120701.1
NM_001127230.1 702 Missense Mutation ATC,ATG I176M NP_001120702.1
NM_017900.2 702 Missense Mutation ATC,ATG I176M NP_060370.1
XM_011541670.2 702 Missense Mutation ATC,ATG I205M XP_011539972.1

View Full Product Details