Product Details
- SNP ID
-
rs138176961
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:37683354 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTTGGTGTTCGTCTTTTGAAATT[C/T]GGTCCCAGGCCTTGGGCAAAGCTTG
- Phenotype
-
MIM: 614799
MIM: 609977
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf109
PubMed Links
Gene Details
- Gene
- C1orf109
- Gene Name
- chromosome 1 open reading frame 109
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001303030.1 |
745 |
Missense Mutation |
CAA,CGA |
Q242R |
NP_001289959.1 |
NM_001303031.1 |
745 |
Missense Mutation |
CAA,CGA |
Q243R |
NP_001289960.1 |
NM_017850.2 |
745 |
Missense Mutation |
CAA,CGA |
Q180R |
NP_060320.1 |
XM_005270975.4 |
745 |
Silent Mutation |
CCA,CCG |
P243P |
XP_005271032.1 |
XM_005270979.1 |
745 |
Silent Mutation |
CCA,CCG |
P181P |
XP_005271036.1 |
XM_005270980.1 |
745 |
Silent Mutation |
CCA,CCG |
P181P |
XP_005271037.1 |
XM_005270981.4 |
745 |
Silent Mutation |
CCA,CCG |
P181P |
XP_005271038.1 |
XM_005270982.1 |
745 |
Silent Mutation |
CCA,CCG |
P181P |
XP_005271039.1 |
XM_011541637.1 |
745 |
Silent Mutation |
CCA,CCG |
P244P |
XP_011539939.1 |
XM_011541638.1 |
745 |
Silent Mutation |
CCA,CCG |
P244P |
XP_011539940.1 |
XM_011541639.1 |
745 |
Silent Mutation |
CCA,CCG |
P243P |
XP_011539941.1 |
XM_011541640.1 |
745 |
Silent Mutation |
CCA,CCG |
P181P |
XP_011539942.1 |
XM_011541641.1 |
745 |
Silent Mutation |
CCA,CCG |
P181P |
XP_011539943.1 |
XM_011541642.1 |
745 |
Silent Mutation |
CCA,CCG |
P181P |
XP_011539944.1 |
XM_011541643.1 |
745 |
Silent Mutation |
CCA,CCG |
P181P |
XP_011539945.1 |
XM_011541646.2 |
745 |
Silent Mutation |
CCA,CCG |
P244P |
XP_011539948.1 |
XM_017001544.1 |
745 |
Silent Mutation |
CCA,CCG |
P244P |
XP_016857033.1 |
XM_017001545.1 |
745 |
Silent Mutation |
CCA,CCG |
P181P |
XP_016857034.1 |
XM_017001546.1 |
745 |
Missense Mutation |
CAA,CGA |
Q180R |
XP_016857035.1 |
XM_017001547.1 |
745 |
Missense Mutation |
CAA,CGA |
Q180R |
XP_016857036.1 |
XM_017001548.1 |
745 |
Missense Mutation |
CAA,CGA |
Q242R |
XP_016857037.1 |
XM_017001549.1 |
745 |
Missense Mutation |
CAA,CGA |
Q242R |
XP_016857038.1 |
XM_017001550.1 |
745 |
Missense Mutation |
CAA,CGA |
Q243R |
XP_016857039.1 |
XM_017001551.1 |
745 |
Missense Mutation |
CAA,CGA |
Q180R |
XP_016857040.1 |
XM_017001552.1 |
745 |
Missense Mutation |
CAA,CGA |
Q180R |
XP_016857041.1 |
XM_017001553.1 |
745 |
Missense Mutation |
CAA,CGA |
Q180R |
XP_016857042.1 |
XM_017001554.1 |
745 |
Missense Mutation |
CAA,CGA |
Q180R |
XP_016857043.1 |
XM_017001555.1 |
745 |
Missense Mutation |
CAA,CGA |
Q180R |
XP_016857044.1 |
XM_017001556.1 |
745 |
Missense Mutation |
CAA,CGA |
Q180R |
XP_016857045.1 |
- Gene
- CDCA8
- Gene Name
- cell division cycle associated 8
There are no transcripts associated with this gene.
View Full Product Details