Product Details
- SNP ID
-
rs138218533
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:197040576 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACTGCCCTCTGTCACATTGCATT[C/G]TAAGTATAGATCCAGTAATATATAA
- Phenotype
-
MIM: 134580
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
F13B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5998] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- F13B
- Gene Name
- coagulation factor XIII B chain
View Full Product Details