Product Details

SNP ID
rs138220875
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:44823290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAGCAAGCATGAGCAGACCCAG[C/G]AATGTGGAGATGGCCCCATCGGTCA
Phenotype
MIM: 603673
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
There are no transcripts associated with this gene.

Gene
PTCH2
Gene Name
patched 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166292.1 3222 Missense Mutation TTC,TTG F1070L NP_001159764.1
NM_003738.4 3222 Missense Mutation TTC,TTG F1070L NP_003729.3

View Full Product Details