Product Details

SNP ID
rs138290082
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:248038821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTGATGTTCCAGCTATGTTGA[C/T]GCTAGCCTGCACAGACACTTGGGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2L13 PubMed Links

Gene Details

Gene
OR2L13
Gene Name
olfactory receptor family 2 subfamily L member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304535.1 780 Intron NP_001291464.1
NM_175911.3 780 Intron NP_787107.1
XM_011544169.2 780 Intron XP_011542471.1
Gene
OR2L2
Gene Name
olfactory receptor family 2 subfamily L member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004686.2 780 Missense Mutation ACG,ATG T185M NP_001004686.1
XM_011544158.2 780 Missense Mutation ACG,ATG T185M XP_011542460.1

View Full Product Details