Product Details

SNP ID
rs138321348
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:108753123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATCGTGTTGCAAAATGACAGAT[C/G]TTCTAGAAGAAGGTATTACTGGTAA
Phenotype
MIM: 608339
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STXBP3 PubMed Links

Gene Details

Gene
STXBP3
Gene Name
syntaxin binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007269.2 235 Missense Mutation CTT,GTT L54V NP_009200.2

View Full Product Details