Product Details

SNP ID
rs138419867
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:205303485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAGCAGACCCTCAGTGCCTGTC[A/G]GTAGGTCGCTGTCACCTCCTGGCAG
Phenotype
MIM: 608131
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUAK2 PubMed Links

Gene Details

Gene
NUAK2
Gene Name
NUAK family kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030952.2 1272 Nonsense Mutation CGA,TGA R662* NP_112214.2
XM_005245515.4 1272 Nonsense Mutation CGA,TGA R371* XP_005245572.1

View Full Product Details