Product Details

SNP ID
rs138436244
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:54139638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATGGAGAAGGAGCTGGAGAAGA[A/C]CATTCATGGGGGGGGCAGGACAAAC
Phenotype
MIM: 612320
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDCP2 PubMed Links

Gene Details

Gene
CDCP2
Gene Name
CUB domain containing protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201546.3 1316 Missense Mutation NP_963840.2

View Full Product Details