Product Details
- SNP ID
-
rs138449369
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:155950455 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGACAGAGCGAGTAGTGACAGGTA[A/G]ATCCAGGCGGTCAGTGCCTCGGCTG
- Phenotype
-
MIM: 607560
MIM: 609043
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF2
PubMed Links
Gene Details
- Gene
- ARHGEF2
- Gene Name
- Rho/Rac guanine nucleotide exchange factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001162383.1 |
2893 |
Silent Mutation |
CTA,TTA |
L911L |
NP_001155855.1 |
NM_001162384.1 |
2893 |
Silent Mutation |
CTA,TTA |
L910L |
NP_001155856.1 |
NM_004723.3 |
2893 |
Silent Mutation |
CTA,TTA |
L883L |
NP_004714.2 |
XM_005245587.2 |
2893 |
Silent Mutation |
CTA,TTA |
L911L |
XP_005245644.1 |
XM_005245588.2 |
2893 |
Silent Mutation |
CTA,TTA |
L910L |
XP_005245645.1 |
XM_005245589.4 |
2893 |
Silent Mutation |
CTA,TTA |
L903L |
XP_005245646.1 |
XM_005245590.3 |
2893 |
Silent Mutation |
CTA,TTA |
L896L |
XP_005245647.1 |
XM_005245591.3 |
2893 |
Silent Mutation |
CTA,TTA |
L896L |
XP_005245648.1 |
XM_005245592.2 |
2893 |
Silent Mutation |
CTA,TTA |
L884L |
XP_005245649.1 |
XM_005245593.4 |
2893 |
Silent Mutation |
CTA,TTA |
L884L |
XP_005245650.1 |
XM_005245594.2 |
2893 |
Silent Mutation |
CTA,TTA |
L884L |
XP_005245651.1 |
XM_006711622.3 |
2893 |
Silent Mutation |
CTA,TTA |
L894L |
XP_006711685.1 |
XM_006711623.3 |
2893 |
Silent Mutation |
CTA,TTA |
L835L |
XP_006711686.1 |
XM_006711624.3 |
2893 |
Silent Mutation |
CTA,TTA |
L835L |
XP_006711687.1 |
XM_006711625.3 |
2893 |
Silent Mutation |
CTA,TTA |
L835L |
XP_006711688.1 |
XM_006711626.2 |
2893 |
Silent Mutation |
CTA,TTA |
L835L |
XP_006711689.1 |
XM_011510136.2 |
2893 |
Silent Mutation |
CTA,TTA |
L1053L |
XP_011508438.2 |
XM_011510137.2 |
2893 |
Silent Mutation |
CTA,TTA |
L1054L |
XP_011508439.2 |
XM_011510139.2 |
2893 |
Silent Mutation |
CTA,TTA |
L1054L |
XP_011508441.1 |
XM_011510141.2 |
2893 |
Silent Mutation |
CTA,TTA |
L907L |
XP_011508443.1 |
XM_011510143.2 |
2893 |
Silent Mutation |
CTA,TTA |
L732L |
XP_011508445.1 |
XM_017002794.1 |
2893 |
Silent Mutation |
CTA,TTA |
L1053L |
XP_016858283.1 |
XM_017002795.1 |
2893 |
Silent Mutation |
CTA,TTA |
L902L |
XP_016858284.1 |
XM_017002796.1 |
2893 |
Silent Mutation |
CTA,TTA |
L893L |
XP_016858285.1 |
XM_017002797.1 |
2893 |
Silent Mutation |
CTA,TTA |
L883L |
XP_016858286.1 |
XM_017002798.1 |
2893 |
Silent Mutation |
CTA,TTA |
L835L |
XP_016858287.1 |
XM_017002799.1 |
2893 |
Silent Mutation |
CTA,TTA |
L834L |
XP_016858288.1 |
- Gene
- MIR6738
- Gene Name
- microRNA 6738
There are no transcripts associated with this gene.
- Gene
- RXFP4
- Gene Name
- relaxin/insulin like family peptide receptor 4
There are no transcripts associated with this gene.
View Full Product Details