Product Details

SNP ID
rs138453986
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:67001992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGGTGACAGATCTCCTAGGGAA[C/T]CTGACCCCCAAGAGGCAGTTCAACT
Phenotype
MIM: 610804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35D1 PubMed Links

Gene Details

Gene
SLC35D1
Gene Name
solute carrier family 35 member D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015139.2 3881 UTR 3 NP_055954.1
XM_006710478.1 3881 UTR 3 XP_006710541.1
XM_011541070.1 3881 UTR 3 XP_011539372.1

View Full Product Details