Product Details

SNP ID
rs138486317
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:153948322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCTCCTTAGAGAAACCTGGAGA[C/G]GAGTCCCCTGTGGGTAGAAGAGACA
Phenotype
MIM: 608972
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRTC2 PubMed Links

Gene Details

Gene
CRTC2
Gene Name
CREB regulated transcription coactivator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181715.2 2028 Silent Mutation TCC,TCG S623S NP_859066.1
XM_005244946.1 2028 Silent Mutation TCC,TCG S628S XP_005245003.1
XM_005244949.2 2028 Silent Mutation TCC,TCG S515S XP_005245006.1
XM_017000575.1 2028 Silent Mutation TCC,TCG S550S XP_016856064.1
XM_017000576.1 2028 Silent Mutation TCC,TCG S545S XP_016856065.1
XM_017000577.1 2028 Silent Mutation TCC,TCG S527S XP_016856066.1
XM_017000578.1 2028 Silent Mutation TCC,TCG S522S XP_016856067.1
XM_017000579.1 2028 Silent Mutation TCC,TCG S417S XP_016856068.1
Gene
DENND4B
Gene Name
DENN domain containing 4B
There are no transcripts associated with this gene.

View Full Product Details