Product Details

SNP ID
rs138487080
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:153991174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAAGAGGAAACACAAGAAGAA[A/G]CGCCTGGTGCAGAGCCCCAATTCCT
Phenotype
MIM: 602672 MIM: 603702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUP210L PubMed Links

Gene Details

Gene
NUP210L
Gene Name
nucleoporin 210 like
There are no transcripts associated with this gene.

Gene
RAB13
Gene Name
RAB13, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272038.1 100 Intron NP_001258967.1
NM_002870.3 100 Intron NP_002861.1
XM_017001959.1 100 Intron XP_016857448.1
Gene
RPS27
Gene Name
ribosomal protein S27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030.4 100 Silent Mutation NP_001021.1

View Full Product Details