Product Details

SNP ID
rs138562827
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:150646516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACACATGGACTGGCTTTTGTCCT[A/G]TTAGAGAGCCAGACTGAAGCTGTAT
Phenotype
MIM: 612208
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLPH3L PubMed Links

Gene Details

Gene
GOLPH3L
Gene Name
golgi phosphoprotein 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018178.5 2807 UTR 3 NP_060648.2
XM_006711428.1 2807 UTR 3 XP_006711491.1

View Full Product Details