Product Details
- SNP ID
-
rs138588851
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:158717346 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAGTGTCGCTGAAACGCAAGTAC[A/C]TGAGTGATACACTGCCAAAGAATAT
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR6K3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28568406] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR6K3
- Gene Name
- olfactory receptor family 6 subfamily K member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005327.2 |
770 |
Missense Mutation |
AGG,ATG |
R257M |
NP_001005327.2 |
View Full Product Details