Product Details

SNP ID
rs138614766
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:159829916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCAAGTGCTGAGCAAAGTCGGG[A/G]GCTCGGTGCTGCTGGTGGCAGCGCG
Phenotype
MIM: 606620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf204 PubMed Links

Gene Details

Gene
C1orf204
Gene Name
chromosome 1 open reading frame 204
There are no transcripts associated with this gene.

Gene
SLAMF8
Gene Name
SLAM family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020125.2 340 Missense Mutation AGC,GGC S31G NP_064510.1
XM_005245345.2 340 Missense Mutation AGC,GGC S31G XP_005245402.1
XM_011509775.1 340 Missense Mutation AGC,GGC S31G XP_011508077.1
XM_011509776.2 340 Missense Mutation AGC,GGC S25G XP_011508078.1
XM_011509777.2 340 Missense Mutation AGC,GGC S31G XP_011508079.1
XM_011509778.2 340 Intron XP_011508080.1
XM_011509779.2 340 Intron XP_011508081.1
XM_011509780.2 340 Intron XP_011508082.2
XM_017001844.1 340 Missense Mutation AGC,GGC S31G XP_016857333.1
XM_017001845.1 340 Missense Mutation AGC,GGC S31G XP_016857334.1
XM_017001846.1 340 Missense Mutation AGC,GGC S25G XP_016857335.1

View Full Product Details