Product Details

SNP ID
rs138656008
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:37567741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCTCTTCCCTTTGTGCCTTCC[A/G]CTTCTTTCCCTTTTTGGAAGGTGCT
Phenotype
MIM: 602135 MIM: 609365
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNALI1 PubMed Links

Gene Details

Gene
DNALI1
Gene Name
dynein axonemal light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003462.3 2138 Intron NP_003453.2
XM_005271172.2 2138 Intron XP_005271229.1
Gene
GNL2
Gene Name
G protein nucleolar 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323623.1 2138 Missense Mutation CGG,TGG R727W NP_001310552.1
NM_001323624.1 2138 Missense Mutation CGG,TGG R476W NP_001310553.1
NM_013285.2 2138 Missense Mutation CGG,TGG R659W NP_037417.1

View Full Product Details