Product Details

SNP ID
rs138700033
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:46184262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGCATCCTTTCTGAAGATCTT[C/T]GGGCCACTGTCGTCCAGTGCCATGC
Phenotype
MIM: 606076 MIM: 606822 MIM: 613170
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIK3R3 PubMed Links

Gene Details

Gene
PIK3R3
Gene Name
phosphoinositide-3-kinase regulatory subunit 3
There are no transcripts associated with this gene.

Gene
POMGNT1
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
There are no transcripts associated with this gene.

Gene
TSPAN1
Gene Name
tetraspanin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005727.3 577 Silent Mutation TTC,TTT F43F NP_005718.2
XM_011540460.2 577 Silent Mutation TTC,TTT F43F XP_011538762.1

View Full Product Details