Product Details

SNP ID
rs138744711
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:24360881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCAGGCCTGGCTGAGGCGGCG[C/T]CGCTGTCCACCGGCTGGTATTGGAC
Phenotype
MIM: 608317 MIM: 615826
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRHL3 PubMed Links

Gene Details

Gene
GRHL3
Gene Name
grainyhead like transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195010.1 1284 Intron NP_001181939.1
NM_021180.3 1284 Intron NP_067003.2
NM_198173.2 1284 Intron NP_937816.1
NM_198174.2 1284 Intron NP_937817.3
XM_011541869.1 1284 Intron XP_011540171.1
XM_011541870.2 1284 Intron XP_011540172.1
Gene
STPG1
Gene Name
sperm tail PG-rich repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199012.1 1284 Missense Mutation NP_001185941.1
NM_001199013.1 1284 Missense Mutation NP_001185942.1
NM_001199014.1 1284 Missense Mutation NP_001185943.1
NM_178122.4 1284 Missense Mutation NP_835223.1
XM_006711025.2 1284 Missense Mutation XP_006711088.1
XM_011542403.2 1284 Missense Mutation XP_011540705.1
XM_011542404.1 1284 Missense Mutation XP_011540706.1
XM_017002768.1 1284 Missense Mutation XP_016858257.1
XM_017002769.1 1284 Missense Mutation XP_016858258.1
XM_017002770.1 1284 Missense Mutation XP_016858259.1

View Full Product Details