Product Details

SNP ID
rs138788038
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:36455833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTGGTATGGCTTTGGCAGGGCT[A/G]TCTGGGTTCCTCCGCTTTGCTTGTT
Phenotype
MIM: 611979 MIM: 608854
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS15 PubMed Links

Gene Details

Gene
MRPS15
Gene Name
mitochondrial ribosomal protein S15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031280.3 893 Silent Mutation GAC,GAT D243D NP_112570.2
Gene
OSCP1
Gene Name
organic solute carrier partner 1
There are no transcripts associated with this gene.

View Full Product Details