Product Details

SNP ID
rs138936363
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:248406340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCATGTACTTCCTCCTCAGCCAC[C/G]TTTCCTTAATTGACATGATGTATAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR2T1 PubMed Links

Gene Details

Gene
OR2T1
Gene Name
olfactory receptor family 2 subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030904.1 346 Missense Mutation CTT,GTT L116V NP_112166.1

View Full Product Details