Product Details

SNP ID
rs138994589
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:66813242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAATGCTTTATAGAACAGCCCAT[C/T]ATGTCCTGTGGCCTCAGTTTAACCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR78 PubMed Links

Gene Details

Gene
WDR78
Gene Name
WD repeat domain 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024763.4 3249 UTR 3 NP_079039.4
NM_207014.2 3249 Intron NP_996897.2
XM_005271212.3 3249 UTR 3 XP_005271269.1
XM_011542161.2 3249 UTR 3 XP_011540463.1
XM_011542162.2 3249 Intron XP_011540464.1
XM_017002353.1 3249 UTR 3 XP_016857842.1
XM_017002354.1 3249 UTR 3 XP_016857843.1
XM_017002355.1 3249 UTR 3 XP_016857844.1
XM_017002356.1 3249 UTR 3 XP_016857845.1
XM_017002357.1 3249 Intron XP_016857846.1

View Full Product Details