Product Details

SNP ID
rs139038197
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:221702531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGTTAAGGTTTGGGGAGATAA[C/T]TGGTCGTTTGCCTTTGACAAATTTA
Phenotype
MIM: 608867
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP10 PubMed Links

Gene Details

Gene
DUSP10
Gene Name
dual specificity phosphatase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007207.5 1569 Missense Mutation NP_009138.1
XM_017000147.1 1569 Intron XP_016855636.1

View Full Product Details