Product Details

SNP ID
rs139044055
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:173803072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATGGAAATGTGAATAAAGGCAA[C/T]CCATGAATAGGTCAACTTCTTCCTG
Phenotype
MIM: 611503
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPL PubMed Links

Gene Details

Gene
CENPL
Gene Name
centromere protein L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127181.2 1457 Missense Mutation GAT,GGT D331G NP_001120653.1
NM_001171182.1 1457 Missense Mutation GAT,GGT D285G NP_001164653.1
NM_033319.3 1457 Missense Mutation GAT,GGT D285G NP_201576.1

View Full Product Details