Product Details

SNP ID
rs139074238
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:109497615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCCCTAGATCATGGCAAGGCCT[A/G]ACAACAGCTGAGCCAGGAAAGTGCT
Phenotype
MIM: 615689
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMIGO1 PubMed Links

Gene Details

Gene
AMIGO1
Gene Name
adhesion molecule with Ig like domain 1
There are no transcripts associated with this gene.

Gene
ATXN7L2
Gene Name
ataxin 7 like 2
There are no transcripts associated with this gene.

Gene
CYB561D1
Gene Name
cytochrome b561 family member D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134400.1 3300 UTR 3 NP_001127872.1
NM_001134402.1 3300 UTR 3 NP_001127874.1
NM_001134403.1 3300 UTR 3 NP_001127875.1
NM_001134404.1 3300 UTR 3 NP_001127876.1
NM_182580.2 3300 UTR 3 NP_872386.1
XM_005270775.2 3300 UTR 3 XP_005270832.1
XM_005270776.3 3300 UTR 3 XP_005270833.1
XM_005270777.2 3300 UTR 3 XP_005270834.1
XM_011541286.2 3300 UTR 3 XP_011539588.1
XM_011541287.2 3300 UTR 3 XP_011539589.2
XM_017001078.1 3300 UTR 3 XP_016856567.1
XM_017001079.1 3300 UTR 3 XP_016856568.1

View Full Product Details