Product Details

SNP ID
rs139081156
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:36097872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCCAGTGGCTGGGTTGTAGCC[A/G]CTGTGGCCATTGTAGAGAGTCCGGT
Phenotype
MIM: 610624 MIM: 120252 MIM: 608953
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADPRHL2 PubMed Links

Gene Details

Gene
ADPRHL2
Gene Name
ADP-ribosylhydrolase like 2
There are no transcripts associated with this gene.

Gene
COL8A2
Gene Name
collagen type VIII alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294347.1 2598 Silent Mutation NP_001281276.1
NM_005202.3 2598 Silent Mutation NP_005193.1
XM_005270477.3 2598 Silent Mutation XP_005270534.1
Gene
TEKT2
Gene Name
tektin 2
There are no transcripts associated with this gene.

View Full Product Details