Product Details

SNP ID
rs139089532
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:205094896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGTCAAGGGCTGGCTGAGTTCC[A/G]CCTGCACTTTACCCTTCGCTGATCC
Phenotype
MIM: 600697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBBP5 PubMed Links

Gene Details

Gene
RBBP5
Gene Name
RB binding protein 5, histone lysine methyltransferase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193272.1 1712 Intron NP_001180201.1
NM_001193273.1 1712 Missense Mutation GCG,GTG A395V NP_001180202.1
NM_005057.3 1712 Missense Mutation GCG,GTG A522V NP_005048.2
XM_011509833.2 1712 Missense Mutation GCG,GTG A522V XP_011508135.1

View Full Product Details