Product Details

SNP ID
rs139141091
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:183627421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAGATATGAAAAAGCAAGAAGG[C/T]AAAGCTGAGAGAAACAGATGCTACC
Phenotype
MIM: 604227 MIM: 605667
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC5 PubMed Links

Gene Details

Gene
ARPC5
Gene Name
actin related protein 2/3 complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270439.1 849 UTR 3 NP_001257368.1
NM_005717.3 849 UTR 3 NP_005708.1
Gene
RGL1
Gene Name
ral guanine nucleotide dissociation stimulator like 1
There are no transcripts associated with this gene.

View Full Product Details