Product Details
- SNP ID
-
rs139205111
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
10
- Location
-
Chr.1:154273442 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGTGGGAACCCAAGGTTCCATAGT[C/G]CTCAGCACCCCCCTGAGGAATTTGG
- Phenotype
-
MIM: 605998
MIM: 616472
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HAX1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs13796] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HAX1
- Gene Name
- HCLS1 associated protein X-1
- Gene
- UBAP2L
- Gene Name
- ubiquitin associated protein 2 like
There are no transcripts associated with this gene.
View Full Product Details