Product Details

SNP ID
rs139237239
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:192811002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGATGGTCTTGCTGCATTCAGGG[C/G]TTTTTTAAAGTCGGAATTCTGTGAA
Phenotype
MIM: 600861
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RGS2 PubMed Links

Gene Details

Gene
RGS2
Gene Name
regulator of G-protein signaling 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002923.3 329 Missense Mutation GCT,GGT A99G NP_002914.1

View Full Product Details