Product Details

SNP ID
rs139246652
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:26794135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCCTCTTCAATGCCATCATCC[C/T]AGATCACCATGCAGAAGCCTTCTCT
Phenotype
MIM: 610274
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIGV PubMed Links

Gene Details

Gene
PIGV
Gene Name
phosphatidylinositol glycan anchor biosynthesis class V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202554.1 790 Missense Mutation CCA,CTA P34L NP_001189483.1
NM_017837.3 790 Missense Mutation CCA,CTA P34L NP_060307.2

View Full Product Details