Product Details

SNP ID
rs139254695
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:225886740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACTAGGCGCCTATGGCTGGAGC[C/T]TCCTTGGCACGAGCGCACCATCCGA
Phenotype
MIM: 603037
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LEFTY1 PubMed Links

Gene Details

Gene
LEFTY1
Gene Name
left-right determination factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020997.3 1168 Missense Mutation AAG,AGG K363R NP_066277.1
Gene
TMEM63A
Gene Name
transmembrane protein 63A
There are no transcripts associated with this gene.

View Full Product Details