Product Details

SNP ID
rs139349948
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:151877106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGTTGGCTATTTATCATAACAA[A/C]AGAACAAAGAGGGATAGGTCTGCAG
Phenotype
MIM: 606388
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
THEM4 PubMed Links

Gene Details

Gene
THEM4
Gene Name
thioesterase superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053055.4 1004 Missense Mutation GTT,TTT V193F NP_444283.2
XM_011509142.2 1004 Intron XP_011507444.1

View Full Product Details