Product Details

SNP ID
rs139391660
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:54032222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCATGATCTTCAGTTTTAGA[C/T]CTAACAACCACAAGAGAAGAAGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM59 PubMed Links

Gene Details

Gene
TMEM59
Gene Name
transmembrane protein 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305043.1 823 Silent Mutation AGA,AGG R301R NP_001291972.1
NM_001305049.1 823 Silent Mutation AGA,AGG R170R NP_001291978.1
NM_001305050.1 823 Silent Mutation AGA,AGG R234R NP_001291979.1
NM_001305051.1 823 Silent Mutation AGA,AGG R169R NP_001291980.1
NM_001305052.1 823 Silent Mutation AGA,AGG R169R NP_001291981.1
NM_001305066.1 823 Intron NP_001291995.1
NM_004872.4 823 Silent Mutation AGA,AGG R300R NP_004863.2
XM_006711051.1 823 Silent Mutation AGA,AGG R233R XP_006711114.1

View Full Product Details