Product Details

SNP ID
rs139414061
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:150487516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTACAGGCTTGCAGGCTACACAC[A/G]GTGCGTGAGGCACCCCCAAAGCTCC
Phenotype
MIM: 612805
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6878 PubMed Links

Gene Details

Gene
MIR6878
Gene Name
microRNA 6878
There are no transcripts associated with this gene.

Gene
TARS2
Gene Name
threonyl-tRNA synthetase 2, mitochondrial (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271895.1 116 Silent Mutation ACA,ACG T22T NP_001258824.1
NM_001271896.1 116 Silent Mutation ACA,ACG T22T NP_001258825.1
NM_025150.4 116 Silent Mutation ACA,ACG T22T NP_079426.2
XM_006711555.1 116 Silent Mutation ACA,ACG T22T XP_006711618.1
XM_006711556.1 116 Silent Mutation ACA,ACG T22T XP_006711619.1
XM_017002394.1 116 Silent Mutation ACA,ACG T22T XP_016857883.1
XM_017002395.1 116 Silent Mutation ACA,ACG T22T XP_016857884.1

View Full Product Details