Product Details

SNP ID
rs139527402
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:10947838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTGGGTAGCAACACTCCTGTG[G/T]GAGCCCACAGGCTCAGGGTTTGCGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf127 PubMed Links

Gene Details

Gene
C1orf127
Gene Name
chromosome 1 open reading frame 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170754.1 2215 Missense Mutation CAC,CCC H766P NP_001164225.1
XM_017000373.1 2215 Missense Mutation CAC,CCC H730P XP_016855862.1

View Full Product Details