Product Details

SNP ID
rs139544420
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:42155542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCTCTCCCTGCCCAGGGACCAT[C/T]GCTAACGACGACTGTGAGCTGTGTG
Phenotype
MIM: 139392 MIM: 601271
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCA2A PubMed Links

Gene Details

Gene
GUCA2A
Gene Name
guanylate cyclase activator 2A
There are no transcripts associated with this gene.

Gene
GUCA2B
Gene Name
guanylate cyclase activator 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007102.2 315 Silent Mutation ATC,ATT I95I NP_009033.1

View Full Product Details