Product Details

SNP ID
rs139608588
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:151849513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCACCCAGTTGGACTCCTTGTC[C/T]CACAGCTCACTACATGTTTGATCTT
Phenotype
MIM: 615653
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2CD4D PubMed Links

Gene Details

Gene
C2CD4D
Gene Name
C2 calcium dependent domain containing 4D
There are no transcripts associated with this gene.

Gene
LOC100132111
Gene Name
uncharacterized LOC100132111
There are no transcripts associated with this gene.

Gene
THEM5
Gene Name
thioesterase superfamily member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182578.3 Intron NP_872384.1
XM_011509421.1 Intron XP_011507723.1

View Full Product Details