Product Details

SNP ID
rs139612031
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:25835242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGTCCTTGTCATTCTTTTCATT[A/G]TCCCAGGAAAACACAGAAACTGGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AUNIP PubMed Links

Gene Details

Gene
AUNIP
Gene Name
aurora kinase A and ninein interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287490.1 926 Silent Mutation GAC,GAT D275D NP_001274419.1
NM_024037.2 926 Silent Mutation GAC,GAT D275D NP_076942.1
XM_017002300.1 926 Silent Mutation GAC,GAT D240D XP_016857789.1
Gene
MTFR1L
Gene Name
mitochondrial fission regulator 1 like
There are no transcripts associated with this gene.

View Full Product Details