Product Details

SNP ID
rs139645876
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:152302701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAATTCCTTTGCCATTAATTTCT[C/T]ACTCATAGTAATAGTATCTCTGTGA
Phenotype
MIM: 135940
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLG PubMed Links

Gene Details

Gene
FLG
Gene Name
filaggrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002016.1 12221 Silent Mutation TAA,TGA *4062* NP_002007.1

View Full Product Details