Product Details
- SNP ID
-
rs139649211
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:32274764 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCATCCGAAATGGCTCTGAGGTG[C/T]GGGACCCACTGGTTACCTACGAAGG
- Phenotype
-
MIM: 153390
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LCK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145088108] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LCK
- Gene Name
- LCK proto-oncogene, Src family tyrosine kinase
View Full Product Details