Product Details
- SNP ID
-
rs139660935
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:247860621 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGAAATGTTCCCAAACAGGTAA[A/G]GCTCCAGATGGCTCTGGAACTTATG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRIM58
PubMed Links
Gene Details
- Gene
- TRIM58
- Gene Name
- tripartite motif containing 58
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015431.3 |
473 |
Missense Mutation |
AAG,AGG |
K142R |
NP_056246.3 |
View Full Product Details