Product Details

SNP ID
rs139664603
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:177932764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCAGGGGAAGGTGTCAGTGAGA[A/G]GCCCAGGCCAGCCTGGTGGGGAGAA
Phenotype
MIM: 612855
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC16B PubMed Links

Gene Details

Gene
SEC16B
Gene Name
SEC16 homolog B, endoplasmic reticulum export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033127.2 2956 Missense Mutation CTC,TTC L956F NP_149118.2

View Full Product Details