Product Details

SNP ID
rs139705648
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:35603338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTTTTGTCAATGATTCGAACAC[C/T]GAAGGTTGGCAGATTCAGGATGAAG
Phenotype
MIM: 602175 MIM: 614428
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMB2 PubMed Links

Gene Details

Gene
PSMB2
Gene Name
proteasome subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199779.1 579 Missense Mutation AGT,GGT S154G NP_001186708.1
NM_001199780.1 579 Missense Mutation AGT,GGT S62G NP_001186709.1
NM_002794.4 579 Missense Mutation AGT,GGT S179G NP_002785.1
Gene
TFAP2E
Gene Name
transcription factor AP-2 epsilon
There are no transcripts associated with this gene.

View Full Product Details