Product Details

SNP ID
rs139724134
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:26794424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATTTCTTGTTCTTCATGTTGGC[C/T]GCAGTTGCACTTCATGACCTGGGTT
Phenotype
MIM: 610274
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIGV PubMed Links

Gene Details

Gene
PIGV
Gene Name
phosphatidylinositol glycan anchor biosynthesis class V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202554.1 1079 Silent Mutation GCC,GCT A130A NP_001189483.1
NM_017837.3 1079 Silent Mutation GCC,GCT A130A NP_060307.2

View Full Product Details