Product Details

SNP ID
rs139834403
Assay Type
Functionally Tested
NCBI dbSNP Submissions
12
Location
Chr.1:248877342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCTTTGCCTAAATGTGGTTTGT[A/G]TATATTGTAGCCAAGCTTGCCGTGG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PGBD2 PubMed Links

Gene Details

Gene
PGBD2
Gene Name
piggyBac transposable element derived 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017434.1 Intron NP_001017434.1
NM_170725.2 Intron NP_733843.1
XM_005270333.1 Intron XP_005270390.1
XM_011544159.2 Intron XP_011542461.1
XM_011544161.2 Intron XP_011542463.1

View Full Product Details