Product Details

SNP ID
rs140004020
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:12242538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTGTTGAATTAGAAAACTTGCC[A/G]TTAAAGAAAGATGCCTTGAAAGAAT
Phenotype
MIM: 608877
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS13D PubMed Links

Gene Details

Gene
VPS13D
Gene Name
vacuolar protein sorting 13 homolog D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015378.3 281 Silent Mutation CCA,CCG P41P NP_056193.2
NM_018156.3 281 Silent Mutation CCA,CCG P41P NP_060626.2

View Full Product Details