Product Details

SNP ID
rs140008766
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:161122453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGCGCTCAAGTGGGTCCTGCTT[A/G]TTAGAGAAGACATTGCCCTGCAGAG
Phenotype
MIM: 606841 MIM: 604618 MIM: 613466
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEDD PubMed Links

Gene Details

Gene
DEDD
Gene Name
death effector domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039711.1 887 Silent Mutation AAC,AAT N217N NP_001034800.1
NM_001039712.1 887 Silent Mutation AAC,AAT N217N NP_001034801.1
NM_032998.2 887 Silent Mutation AAC,AAT N217N NP_127491.1
XM_005245597.3 887 Silent Mutation AAC,AAT N247N XP_005245654.1
XM_005245598.4 887 Silent Mutation AAC,AAT N247N XP_005245655.1
XM_005245599.2 887 Silent Mutation AAC,AAT N247N XP_005245656.1
XM_005245600.2 887 Silent Mutation AAC,AAT N247N XP_005245657.1
XM_017002802.1 887 Silent Mutation AAC,AAT N217N XP_016858291.1
Gene
NIT1
Gene Name
nitrilase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185092.1 887 Intron NP_001172021.1
NM_001185093.1 887 Intron NP_001172022.1
NM_001185094.1 887 Intron NP_001172023.1
NM_005600.2 887 Intron NP_005591.1
XM_005245214.2 887 Intron XP_005245271.1
XM_005245215.4 887 Intron XP_005245272.1
XM_005245216.3 887 Intron XP_005245273.1
XM_006711341.3 887 Intron XP_006711404.1
XM_017001371.1 887 Intron XP_016856860.1
Gene
PFDN2
Gene Name
prefoldin subunit 2
There are no transcripts associated with this gene.

View Full Product Details