Product Details
- SNP ID
-
rs140026272
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:52606753 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGCACTACCGAGCCCTGCAGCAG[C/T]TGCAGCGAGACCTGGGCCCCCACCA
- Phenotype
-
MIM: 615784
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GPX7
PubMed Links
Gene Details
- Gene
- GPX7
- Gene Name
- glutathione peroxidase 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015696.4 |
247 |
Silent Mutation |
CTG,TTG |
L70L |
NP_056511.2 |
View Full Product Details