Product Details

SNP ID
rs140053354
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:95096918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTTGGAAGAAATGAGGGGAGT[C/T]AAAGAAATACAAGTAACTTAGAATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM56 PubMed Links

Gene Details

Gene
TMEM56
Gene Name
transmembrane protein 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199679.1 Intron NP_001186608.1
NM_152487.2 Intron NP_689700.1

View Full Product Details